Assay Details
| Gene Symbol: LRRC47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060460.1 | XP_017915949.1 | 4 - 5 | 2063 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC47| Gene Symbol: LRRC47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060460.1 | XP_017915949.1 | 4 - 5 | 2063 | 60 | |
Set Membership: |
|