Assay Details
| Gene Symbol: CD101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677811.3 | XP_005677868.2 | 8 - 9 | 2900 | 63 | |
| GenBank mRNA | JO332680.2 | - | 8 - 9 | 2883 | 63 | |
More Information
Set Membership: |
|

CD101
Genomic Map