Assay Details
| Gene Symbol: LOC102175263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678505.1 | XP_005678562.1 | 3 - 4 | 469 | 60 | |
| XM_005678506.1 | XP_005678563.1 | 3 - 4 | 469 | 60 | ||
| GenBank mRNA | KA311379.1 | - | 3 - 4 | 478 | 60 | |
More Information
Set Membership: |
|

LOC102175263
Genomic Map