Assay Details
| Gene Symbol: MMAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061101.1 | XP_017916590.1 | 1 - 2 | 204 | 57 | |
| XM_018061102.1 | XP_017916591.1 | 1 - 2 | 205 | 57 | ||
| GenBank mRNA | JP351155.2 | - | 1 - 2 | 222 | 57 | |
| KA320566.1 | - | 1 - 2 | 169 | 57 | ||
More Information
Set Membership: |
|

MMAB
Genomic Map