Assay Details
| Gene Symbol: CXCL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064531.1 | XP_017920020.1 | 5 - 6 | 876 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXCL16| Gene Symbol: CXCL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064531.1 | XP_017920020.1 | 5 - 6 | 876 | 78 | |
Set Membership: |
|