Assay Details
| Gene Symbol: AMZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063943.1 | XP_017919432.1 | 7 - 8 | 1417 | 60 | |
| XM_018063944.1 | XP_017919433.1 | 8 - 9 | 1206 | 60 | ||
| XM_018063945.1 | XP_017919434.1 | 7 - 8 | 1085 | 60 | ||
| GenBank mRNA | JO374563.2 | - | 2 - 3 | 274 | 60 | |
| JO433579.2 | - | 6 - 7 | 1292 | 60 | ||
More Information
Set Membership: |
|

AMZ2
Genomic Map