Assay Details
| Gene Symbol: LOC102178029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696858.3 | XP_005696915.1 | 3 - 4 | 249 | 76 | |
| XM_013974155.2 | XP_013829609.1 | 3 - 4 | 285 | 76 | ||
| GenBank mRNA | JO440522.2 | - | 3 - 4 | 289 | 76 | |
| JO441473.2 | - | 3 - 4 | 244 | 76 | ||
More Information
Set Membership: |
|

LOC102178029
Genomic Map