Assay Details
| Gene Symbol: LOC102178029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696858.3 | XP_005696915.1 | 5 - 6 | 433 | 77 | |
| XM_013974155.2 | XP_013829609.1 | 5 - 6 | 469 | 77 | ||
| GenBank mRNA | JO440522.2 | - | 5 - 6 | 473 | 77 | |
| JO441473.2 | - | 5 - 6 | 428 | 77 | ||
More Information
Set Membership: |
|

LOC102178029
Genomic Map