Assay Details
| Gene Symbol: NSUN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040555.1 | XP_017896044.1 | 6 - 7 | 794 | 59 | |
| GenBank mRNA | KA324802.1 | - | 4 - 5 | 1460 | 59 | |
More Information
Set Membership: |
|

NSUN5
Genomic Map