Assay Details
| Gene Symbol: LOC102173955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059399.1 | XP_017914888.1 | 4 - 5 | 369 | 107 | |
| XM_018059400.1 | XP_017914889.1 | 4 - 5 | 369 | 107 | ||
| XM_018059401.1 | XP_017914890.1 | 4 - 5 | 369 | 107 | ||
More Information
Set Membership: |
|

LOC102173955
Genomic Map