Assay Details
| Gene Symbol: XPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066886.1 | XP_017922375.1 | 6 - 7 | 840 | 64 | |
| GenBank mRNA | JO372135.2 | - | 6 - 7 | 802 | 64 | |
More Information
Set Membership: |
|

XPC
Genomic Map