Assay Details
| Gene Symbol: PGBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974051.2 | XP_013829505.2 | 3 - 4 | 3278 | 73 | |
| XM_013974052.2 | XP_013829506.2 | 4 - 5 | 1587 | 73 | ||
| XM_013974054.2 | XP_013829508.2 | 4 - 5 | 1401 | 73 | ||
More Information
Set Membership: |
|

PGBD1
Genomic Map