Assay Details
| Gene Symbol: BCL7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040874.1 | XP_017896363.1 | 3 - 4 | 632 | 85 | |
| XM_018040875.1 | XP_017896364.1 | 3 - 4 | 632 | 85 | ||
| GenBank mRNA | JO408916.2 | - | 3 - 4 | 376 | 85 | |
More Information
Set Membership: |
|

BCL7C
Genomic Map