Assay Details
| Gene Symbol: S100A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677500.3 | XP_005677557.1 | 2 - 3 | 336 | 102 | |
| GenBank mRNA | JO518240.2 | - | 3 - 4 | 393 | 102 | |
More Information
Set Membership: |
|

S100A2
Genomic Map