Assay Details
| Gene Symbol: SLC25A30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687432.3 | XP_005687489.1 | 2 - 3 | 648 | 60 | |
| XM_018056765.1 | XP_017912254.1 | 2 - 3 | 232 | 60 | ||
More Information
Set Membership: |
|

SLC25A30
Genomic Map