Assay Details
| Gene Symbol: ALG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687612.3 | XP_005687669.2 | 2 - 3 | 307 | 66 | |
| XM_018056662.1 | XP_017912151.1 | 2 - 3 | 289 | 66 | ||
| GenBank mRNA | JO440187.2 | - | 8 - 9 | 826 | 66 | |
| JO540550.2 | - | 4 - 5 | 307 | 66 | ||
More Information
Set Membership: |
|

ALG5
Genomic Map