Assay Details
| Gene Symbol: MED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693626.3 | XP_005693683.1 | 1 - 2 | 278 | 58 | |
| GenBank mRNA | JO646273.2 | - | 1 - 2 | 273 | 58 | |
| JT711789.1 | - | 5 - 6 | 1430 | 58 | ||
More Information
Set Membership: |
|

MED9
Genomic Map