Assay Details
| Gene Symbol: MCEE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695172.3 | XP_005695229.2 | 1 - 2 | 128 | 69 | |
| GenBank mRNA | JO446150.2 | - | 2 - 3 | 720 | 69 | |
| KA306556.1 | - | 2 - 3 | 785 | 69 | ||
More Information
Set Membership: |
|

MCEE
Genomic Map