Assay Details
| Gene Symbol: UBFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040769.1 | XP_017896258.1 | 6 - 7 | 1085 | 79 | |
| GenBank mRNA | JO565848.2 | - | 5 - 6 | 737 | 79 | |
More Information
Set Membership: |
|

UBFD1
Genomic Map