Assay Details
| Gene Symbol: YJEFN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051499.1 | XP_017906988.1 | 4 - 5 | 917 | 53 | |
| XM_018051500.1 | XP_017906989.1 | 5 - 6 | 1060 | 53 | ||
| GenBank mRNA | JO595500.2 | - | 4 - 5 | 475 | 53 | |
More Information
Set Membership: |
|

YJEFN3
Genomic Map