Assay Details
| Gene Symbol: TBX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061190.1 | XP_017916679.1 | 7 - 8 | 5246 | 58 | |
| XM_018061191.1 | XP_017916680.1 | 6 - 7 | 2700 | 58 | ||
More Information
Set Membership: |
|

TBX3
Genomic Map