Assay Details
| Gene Symbol: NLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064292.1 | XP_017919781.1 | 6 - 7 | 2204 | 62 | |
| GenBank mRNA | KA342067.1 | - | 6 - 7 | 984 | 62 | |
More Information
Set Membership: |
|

NLK
Genomic Map