Assay Details
| Gene Symbol: DBF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064012.1 | XP_017919501.1 | 8 - 9 | 753 | 56 | |
| GenBank mRNA | JO582856.1 | - | 2 - 3 | 220 | 56 | |
| JT712194.1 | - | 3 - 4 | 176 | 56 | ||
More Information
Set Membership: |
|

DBF4B
Genomic Map