Assay Details
| Gene Symbol: SLC16A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064517.1 | XP_017920006.1 | 2 - 3 | 349 | 108 | |
| XM_018064518.1 | XP_017920007.1 | 2 - 3 | 344 | 108 | ||
| GenBank mRNA | JO433852.2 | - | 2 - 3 | 384 | 108 | |
| JO446848.2 | - | 2 - 3 | 384 | 108 | ||
More Information
Set Membership: |
|

SLC16A11
Genomic Map