Assay Details
| Gene Symbol: SLC16A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064517.1 | XP_017920006.1 | 4 - 5 | 1266 | 97 | |
| XM_018064518.1 | XP_017920007.1 | 4 - 5 | 1261 | 97 | ||
| XM_018064519.1 | XP_017920008.1 | 4 - 5 | 1291 | 97 | ||
| GenBank mRNA | JO433852.2 | - | 4 - 5 | 1301 | 97 | |
More Information
Set Membership: |
|

SLC16A11
Genomic Map