Assay Details
| Gene Symbol: GPR62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066922.1 | XP_017922411.1 | 4 - 5 | 477 | 68 | |
| GenBank mRNA | JO636447.2 | - | 1 - 2 | 163 | 68 | |
More Information
Set Membership: |
|

GPR62
Genomic Map