Assay Details
| Gene Symbol: SLC41A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066953.1 | XP_017922442.1 | 5 - 6 | 721 | 60 | |
| XM_018066954.1 | XP_017922443.1 | 5 - 6 | 2161 | 60 | ||
| XM_018066955.1 | XP_017922444.1 | 5 - 6 | 751 | 60 | ||
| XM_018066956.1 | XP_017922445.1 | 4 - 5 | 1908 | 60 | ||
| XM_018066957.1 | XP_017922446.1 | 4 - 5 | 4613 | 60 | ||
| GenBank mRNA | JO634205.2 | - | 11 - 12 | 3294 | 60 | |
More Information
Set Membership: |
|

SLC41A3
Genomic Map