Assay Details
| Gene Symbol: TRIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041172.1 | XP_017896661.1 | 2 - 3 | 1367 | 75 | |
| GenBank mRNA | JO635223.2 | - | 2 - 3 | 1444 | 75 | |
| KA315537.1 | - | 2 - 3 | 917 | 75 | ||
More Information
Set Membership: |
|

TRIM8
Genomic Map