Assay Details
| Gene Symbol: GYPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676274.3 | XP_005676331.1 | 3 - 4 | 516 | 75 | |
| XM_005676275.3 | XP_005676332.1 | 2 - 3 | 471 | 75 | ||
More Information
Set Membership: |
|

GYPC
Genomic Map