Assay Details
| Gene Symbol: ISCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686079.3 | XP_005686136.1 | 2 - 3 | 193 | 67 | |
| XM_005686080.3 | XP_005686137.1 | 2 - 3 | 190 | 67 | ||
| XM_013967305.2 | XP_013822759.1 | 2 - 3 | 193 | 67 | ||
| XM_013967306.2 | XP_013822760.1 | 2 - 3 | 190 | 67 | ||
More Information
Set Membership: |
|

ISCA2
Genomic Map