Assay Details
| Gene Symbol: CDK2AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061356.1 | XP_017916845.1 | 3 - 4 | 615 | 63 | |
| GenBank mRNA | JO438004.2 | - | 1 - 2 | 764 | 63 | |
| JO502210.2 | - | 3 - 4 | 542 | 63 | ||
More Information
Set Membership: |
|

CDK2AP1
Genomic Map