Assay Details
| Gene Symbol: TCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691634.3 | XP_005691691.2 | 5 - 6 | 1265 | 72 | |
| GenBank mRNA | JO425767.1 | - | 4 - 5 | 2524 | 72 | |
| JO555625.2 | - | 4 - 5 | 959 | 72 | ||
More Information
Set Membership: |
|

TCN2
Genomic Map