Assay Details
| Gene Symbol: TREML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696299.2 | XP_005696356.1 | 3 - 4 | 480 | 59 | |
| GenBank mRNA | JO342160.2 | - | 1 - 2 | 105 | 59 | |
| JO342161.2 | - | 1 - 2 | 105 | 59 | ||
More Information
Set Membership: |
|

TREML1
Genomic Map