Assay Details
| Gene Symbol: TREML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696299.2 | XP_005696356.1 | 4 - 5 | 572 | 65 | |
| GenBank mRNA | JO342160.2 | - | 2 - 3 | 197 | 65 | |
| JO342161.2 | - | 2 - 3 | 197 | 65 | ||
More Information
Set Membership: |
|

TREML1
Genomic Map