Assay Details
| Gene Symbol: IMPACT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039765.1 | XP_017895254.1 | 9 - 10 | 848 | 62 | |
| GenBank mRNA | KA333493.1 | - | 2 - 3 | 1237 | 62 | |
More Information
Set Membership: |
|

IMPACT
Genomic Map