Assay Details
| Gene Symbol: KCNK18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698618.2 | XP_005698675.2 | 2 - 3 | 347 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNK18| Gene Symbol: KCNK18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698618.2 | XP_005698675.2 | 2 - 3 | 347 | 104 | |
Set Membership: |
|