Assay Details
| Gene Symbol: ETNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060003.1 | XP_017915492.1 | 7 - 8 | 1035 | 65 | |
| XM_018060004.1 | XP_017915493.1 | 6 - 7 | 622 | 65 | ||
More Information
Set Membership: |
|

ETNK2
Genomic Map