Assay Details
| Gene Symbol: LYRM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064289.1 | XP_017919778.1 | 3 - 4 | 304 | 98 | |
| XM_018064290.1 | XP_017919779.1 | 5 - 6 | 526 | 98 | ||
| XM_018064291.1 | XP_017919780.1 | 4 - 5 | 423 | 98 | ||
| GenBank mRNA | KA310014.1 | - | 1 - 2 | 2128 | 98 | |
More Information
Set Membership: |
|

LYRM9
Genomic Map