Assay Details
| Gene Symbol: E2F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055614.1 | XP_017911103.1 | 3 - 4 | 577 | 85 | |
| GenBank mRNA | JO577050.1 | - | 3 - 4 | 733 | 85 | |
More Information
Set Membership: |
|

E2F2
Genomic Map