Assay Details
| Gene Symbol: NDUFS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678668.2 | XP_005678725.1 | 2 - 3 | 386 | 97 | |
| GenBank mRNA | JO633010.2 | - | 1 - 2 | 235 | 97 | |
| KA335456.1 | - | 3 - 4 | 878 | 97 | ||
More Information
Set Membership: |
|

NDUFS5
Genomic Map