Assay Details
| Gene Symbol: PUS7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680062.3 | XP_005680119.2 | 6 - 7 | 1576 | 90 | |
| XM_005680063.3 | XP_005680120.2 | 6 - 7 | 3487 | 90 | ||
| XM_005680064.3 | XP_005680121.2 | 6 - 7 | 1526 | 90 | ||
| XM_013963857.2 | XP_013819311.2 | 7 - 8 | 3576 | 90 | ||
| GenBank mRNA | JT652276.1 | - | 10 - 11 | 1519 | 90 | |
More Information
Set Membership: |
|

PUS7L
Genomic Map