Assay Details
| Gene Symbol: TBC1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683907.3 | XP_005683964.2 | 11 - 12 | 2554 | 63 | |
| XM_005683910.3 | XP_005683967.2 | 10 - 11 | 2046 | 63 | ||
| XM_005683911.3 | XP_005683968.2 | 6 - 7 | 1369 | 63 | ||
| GenBank mRNA | JO596202.2 | - | 2 - 3 | 296 | 63 | |
More Information
Set Membership: |
|

TBC1D2
Genomic Map