Assay Details
| Gene Symbol: FANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041749.1 | XP_017897238.1 | 5 - 6 | 554 | 62 | |
| XM_018041750.1 | XP_017897239.1 | 5 - 6 | 574 | 62 | ||
More Information
Set Membership: |
|

FANK1
Genomic Map