Assay Details
| Gene Symbol: MAT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699270.3 | XP_005699327.2 | 5 - 6 | 768 | 54 | |
| GenBank mRNA | JO421175.2 | - | 4 - 5 | 3983 | 54 | |
| JO421177.2 | - | 4 - 5 | 3890 | 54 | ||
More Information
Set Membership: |
|

MAT1A
Genomic Map