Assay Details
| Gene Symbol: PLPPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050493.1 | XP_017905982.1 | 3 - 4 | 355 | 77 | |
| XM_018050494.1 | XP_017905983.1 | 2 - 3 | 637 | 77 | ||
More Information
Set Membership: |
|

PLPPR3
Genomic Map