Assay Details
| Gene Symbol: CDKN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051898.1 | XP_017907387.1 | 1 - 2 | 526 | 93 | |
| GenBank mRNA | KA334995.1 | - | 1 - 2 | 512 | 93 | |
More Information
Set Membership: |
|

CDKN2B
Genomic Map