Assay Details
| Gene Symbol: LOC106503057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045030.1 | XP_017900519.1 | 3005 | 87 | ||
| XM_018045031.1 | XP_017900520.1 | 3005 | 87 | |||
| Gene Symbol: LOC102182160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060891.1 | XP_017916380.1 | 3017 | 87 | ||
| Gene Symbol: LOC108634983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045299.1 | XP_017900788.1 | 1632 | 87 | ||
More Information
Set Membership: |
|

LOC106503057
Genomic Map