Assay Details
| Gene Symbol: RAB40C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039963.1 | XP_017895452.1 | 4 - 5 | 695 | 69 | |
| XM_018039964.1 | XP_017895453.1 | 3 - 4 | 472 | 69 | ||
| XM_018039965.1 | XP_017895454.1 | 4 - 5 | 479 | 69 | ||
| XM_018039966.1 | XP_017895455.1 | 3 - 4 | 493 | 69 | ||
| GenBank mRNA | JT685931.1 | - | 3 - 4 | 358 | 69 | |
More Information
Set Membership: |
|

RAB40C
Genomic Map