Assay Details
| Gene Symbol: TMEM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679718.3 | XP_005679775.1 | 4 - 5 | 1126 | 70 | |
| XM_005679719.3 | XP_005679776.1 | 4 - 5 | 1126 | 70 | ||
| XM_013963638.2 | XP_013819092.1 | 3 - 4 | 772 | 70 | ||
| GenBank mRNA | JO427458.1 | - | 2 - 3 | 1836 | 70 | |
More Information
Set Membership: |
|

TMEM19
Genomic Map