Assay Details
| Gene Symbol: PRICKLE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680143.3 | XP_005680200.1 | 5 - 6 | 1112 | 63 | |
| XM_018047905.1 | XP_017903394.1 | 8 - 9 | 1155 | 63 | ||
| XM_018047906.1 | XP_017903395.1 | 6 - 7 | 871 | 63 | ||
| XM_018047907.1 | XP_017903396.1 | 6 - 7 | 960 | 63 | ||
| XM_018047908.1 | XP_017903397.1 | 5 - 6 | 1245 | 63 | ||
| XM_018047909.1 | XP_017903398.1 | 5 - 6 | 856 | 63 | ||
More Information
Set Membership: |
|

PRICKLE1
Genomic Map